Please login to your online account to display your discounted pricing

anti-HDAC9 (p Ser155), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224454SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224454SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9734
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish.
Formulation 50μg in 100μL PBS containing 0.05% BSA
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Rabbit
Primary or Secondary Primary
Quantity 0.025mg
Format Affinity Purified
Antigen HDAC9 (p Ser155)
Immunogen A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen.
Regulatory Status RUO
Applications Western Blotting