Please login to your online account to display your discounted pricing

anti-HDAC9 (p Ser155), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224454

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224454

  • / Each
 

Description & Specifications

Specifications

Antigen HDAC9 (p Ser155)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 50μg in 100μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen.
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9734
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish.
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat