Please login to your online account to display your discounted pricing

anti-HDAC7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203974

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203974

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51564
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Test Specificity Cross reacts with Chimpanzee, Cow, Dog, Horse, Human, New World monkey, Mouse and Rat. The amino acid sequence used as immunogen is 100% homologous in human, monkey, cow, dog, chimpanzee and horse and 90% homologous in mouse.
Primary or Secondary Primary
Gene Accession No. NP_060191
Format Affinity Purified
Regulatory Status RUO
Applications Western Blotting
Immunogen Within the range of amino acids 443-463 of HDAC-7 protein were used as the immunogen.
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Quantity 0.1mg
Antigen HDAC7