Please login to your online account to display your discounted pricing

anti-HDAC7, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 443-463 of HDAC-7 protein were used as the immunogen.
Antigen HDAC7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203974SS

 
novus biologicals canada
NBP203974SS
Unlabeled 0.025mg Each for $139.00

NBP203974

 
novus biologicals canada
NBP203974
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 443-463 of HDAC-7 protein were used as the immunogen.
Antigen HDAC7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross reacts with Chimpanzee, Cow, Dog, Horse, Human, New World monkey, Mouse and Rat. The amino acid sequence used as immunogen is 100% homologous in human, monkey, cow, dog, chimpanzee and horse and 90% homologous in mouse.
Gene Accession No. NP_060191
Gene ID (Entrez) 51564
Formulation 100μg in 200μL PBS containing 0.05% BSA