Please login to your online account to display your discounted pricing

anti-HDAC7 (p Ser318), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203978

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203978

  • / Each
 

Description & Specifications

Specifications

Antigen HDAC7 (p Ser318)
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Zebrafish
Format Affinity Purified
Formulation PBS
Gene Accession No. NP_055522
Host Species Rabbit
Immunogen A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen.
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51564
Test Specificity Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Zebrafish. Predicted to react with Mouse, Rat and Chicken.
Target Species Bovine
Target Species Canine
Target Species Equine
Target Species Human
Target Species Primate
Target Species Zebrafish