Please login to your online account to display your discounted pricing

anti-HDAC7 (p Ser318), Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen.
Antigen HDAC7 (p Ser318)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203978SS

 
novus biologicals canada
NBP203978SS
Unlabeled 0.025mg Each for $139.00

NBP203978

 
novus biologicals canada
NBP203978
Unlabeled 0.05mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen.
Antigen HDAC7 (p Ser318)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Zebrafish. Predicted to react with Mouse, Rat and Chicken.
Gene Accession No. NP_055522
Gene ID (Entrez) 51564
Formulation PBS
Concentration 1.0mg/mL