Please login to your online account to display your discounted pricing

anti-HDAC6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178981SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178981SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10013
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9UBN7
Test Specificity Human and mouse. Mouse data from customer feedback.
Concentration 1.07mg/mL
Format Affinity Purified
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Formulation PBS, 30% glycerol
Immunogen A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215) [UniProt Q9UBN7]
Host Species Rabbit
Antigen HDAC6
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Quantity 0.025mL