missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HDAC6, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215) [UniProt Q9UBN7]
Antigen HDAC6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178981 Novus Biologicals Canada
NBP178981
0.1mL Each for $309.05
NBP178981SS Novus Biologicals Canada
NBP178981SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
HDAC6
Affinity Purified
RUO
Human and mouse. Mouse data from customer feedback.
10013
1.07mg/mL
A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215) [UniProt Q9UBN7]
Polyclonal
Primary
Flow Cytometry,Immunocytochemistry,Immunofluorescence
Q9UBN7
PBS, 30% glycerol