missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HDAC5, Polyclonal, Novus Biologicals™

$142.00 - $409.37

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC5 around the phosphorylation site of serine 498 (T-Q-SP-S-P).
Antigen HDAC5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203989 Novus Biologicals Canada
NBP203989
0.1mg Each for $409.37
NBP203989SS Novus Biologicals Canada
NBP203989SS
0.025mg Each for $142.00
Specifications Description & Specifications

Specifications

Rabbit
HDAC5
Affinity Purified
RUO
Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide affinity columns. Rat reactivity reported in scientific literature (PMID: 18292392)
10014
This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC5 around the phosphorylation site of serine 498 (T-Q-SP-S-P).
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
NP_001015053
PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol.