Please login to your online account to display your discounted pricing

anti-HDAC5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203989SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203989SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10014
Format Affinity Purified
Gene Accession No. NP_001015053
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC5 around the phosphorylation site of serine 498 (T-Q-SP-S-P).
Host Species Rabbit
Conjugate Unlabeled
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol.
Test Specificity Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide affinity columns. Rat reactivity reported in scientific literature (PMID: 18292392)
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen HDAC5
Quantity 0.025mg
Primary or Secondary Primary