Please login to your online account to display your discounted pricing

anti-HDAC5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203989

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203989

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10014
Quantity 0.1mg
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide affinity columns. Rat reactivity reported in scientific literature (PMID: 18292392)
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol.
Gene Accession No. NP_001015053
Format Affinity Purified
Host Species Rabbit
Antigen HDAC5
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC5 around the phosphorylation site of serine 498 (T-Q-SP-S-P).