Please login to your online account to display your discounted pricing

anti-HDAC3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001669

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001669

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8841
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Primary or Secondary Primary
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity In HeLa, a 50 kDa band is observed. Cross-reacts with Human. Expected to cross-react with Mouse (100% identity with immunogen), Rat (100% identity with immunogen) and Chicken (100% identity with immunogen) due to sequence homology. Not yet tested in other species.
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 2-17 of human HDAC3.
Antigen HDAC3
Host Species Rabbit
Cross Reactivity Human
Quantity 0.1mg
Regulatory Status RUO
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Purified