Please login to your online account to display your discounted pricing

anti-HDAC3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001669SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001669SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8841
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 2-17 of human HDAC3.
Conjugate Unlabeled
Test Specificity In HeLa, a 50 kDa band is observed. Cross-reacts with Human. Expected to cross-react with Mouse (100% identity with immunogen), Rat (100% identity with immunogen) and Chicken (100% identity with immunogen) due to sequence homology. Not yet tested in other species.
Concentration 0.5mg/mL
Cross Reactivity Human
Host Species Rabbit
Antigen HDAC3
Regulatory Status RUO
Quantity 0.025mg
Primary or Secondary Primary
Format Purified