Please login to your online account to display your discounted pricing

anti-HDAC1, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen 433-448 and 467-482 of human HDAC1 (Genbank Accession no. Q13547).
Antigen HDAC1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056340

 
novus biologicals canada
NB10056340
Unlabeled 0.1mg Each for $362.00

NB10056340T

 
novus biologicals canada
NB10056340SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 433-448 and 467-482 of human HDAC1 (Genbank Accession no. Q13547).
Antigen HDAC1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Test Specificity In 293, a 60 kDa band is observed.
Gene Accession No. Q01101
Gene ID (Entrez) 3065
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL