Please login to your online account to display your discounted pricing

anti-HAPLN3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224476SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224476SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 145864
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), chimpanzee and monkey, 88% homologous in cow, 82% homologous in mouse and rat and 76% homologous in zebrafish.
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Format Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Primate
Antigen HAPLN3
Immunogen A portion of amino acids 1-50 of human HAPLN3 was used as the immunogen.