Please login to your online account to display your discounted pricing

anti-HAPLN3, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human HAPLN3 was used as the immunogen.
Antigen HAPLN3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224476

 
novus biologicals canada
NBP224476
Unlabeled 0.1mg Each for $255.80

NBP224476SS

 
novus biologicals canada
NBP224476SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human HAPLN3 was used as the immunogen.
Antigen HAPLN3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), chimpanzee and monkey, 88% homologous in cow, 82% homologous in mouse and rat and 76% homologous in zebrafish.
Gene ID (Entrez) 145864
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL