Please login to your online account to display your discounted pricing

anti-HA95/AKAP8L, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179140

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP179140

  • / Each
 

Description & Specifications

Specifications

Antigen HA95/AKAP8L
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Gene Accession No. Q9ULX6
Host Species Rabbit
Immunogen Human HA95/AKAP8L peptide within amino acids 50 to 100 [Swiss-Prot# Q9ULX6].
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 26993
Target Species Human