Please login to your online account to display your discounted pricing

anti-HA95/AKAP8L, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Human HA95/AKAP8L peptide within amino acids 50 to 100 [Swiss-Prot# Q9ULX6].
Antigen HA95/AKAP8L
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP179140

 
novus biologicals canada
NBP179140
Unlabeled 0.1mL Each for $309.05

NBP179140SS

 
novus biologicals canada
NBP179140SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Human HA95/AKAP8L peptide within amino acids 50 to 100 [Swiss-Prot# Q9ULX6].
Antigen HA95/AKAP8L
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. Q9ULX6
Gene ID (Entrez) 26993