Please login to your online account to display your discounted pricing

anti-HA95/AKAP8L, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179140SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP179140SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 26993
Gene Accession No. Q9ULX6
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Primary or Secondary Primary
Format Antiserum
Antigen HA95/AKAP8L
Regulatory Status RUO
Immunogen Human HA95/AKAP8L peptide within amino acids 50 to 100 [Swiss-Prot# Q9ULX6].
Cross Reactivity Human