Please login to your online account to display your discounted pricing

anti-HA Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6003631MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6003631MG

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate Buffered Saline (PBS)
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Concentration 1.0mg/mL
Primary or Secondary Primary
Immunogen Rabbits were immunized with HA cleavage site (YPYDVPDYA) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Non-species specific
Antigen HA Epitope Tag
Host Species Rabbit
Format Affinity Purified
Regulatory Status RUO
Quantity 1mg