Please login to your online account to display your discounted pricing

anti-HA Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600363

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600363

  • / Each
 

Description & Specifications

Specifications

Concentration 1.0mg/mL
Cross Reactivity Non-species specific
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg
Primary or Secondary Primary
Conjugate Unlabeled
Formulation phosphate Buffered Saline (PBS)
Format Affinity Purified
Immunogen Rabbits were immunized with HA cleavage site (YPYDVPDYA) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Host Species Rabbit
Antigen HA Epitope Tag