Please login to your online account to display your discounted pricing

anti-H2AX, Polyclonal, Novus Biologicals™

$142.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-153 of human Histone H2A.x was used as the immunogen for the antibody.
Antigen H2AX
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227427SS

 
novus biologicals canada
NBP227427SS
Unlabeled 0.025mg Each for $142.00

NBP227427

 
novus biologicals canada
NBP227427
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-153 of human Histone H2A.x was used as the immunogen for the antibody.
Antigen H2AX
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the Histone H2A.x antibody is 100% homologous in human and monkey, 94% homologous in cow and dog, 88% homologous in rat and 82% homologous in mouse.
Gene Accession No. P16104
Gene ID (Entrez) 3014
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL