Please login to your online account to display your discounted pricing

anti-GST Epitope Tag, Polyclonal, Novus Biologicals™

$274.00 - $633.32

Specifications

Host Species Chicken
Immunogen Chickens were immunized with Glutathione-S- Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Antigen GST Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600327

 
novus biologicals canada
NB600327
Unlabeled 0.1mg Each for $274.00

NB6003271MG

 
novus biologicals canada
NB6003271MG
Unlabeled 1mg Each for $633.32
Description & Specifications

Specifications

Host Species Chicken
Immunogen Chickens were immunized with Glutathione-S- Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Antigen GST Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene ID (Entrez) 2944
Formulation phosphate Buffered Saline (PBS)