Please login to your online account to display your discounted pricing

anti-GST Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6003271MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6003271MG

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2944
Format Affinity Purified
Formulation phosphate Buffered Saline (PBS)
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Antigen GST Epitope Tag
Conjugate Unlabeled
Immunogen Chickens were immunized with Glutathione-S- Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Host Species Chicken
Cross Reactivity Non-species specific
Primary or Secondary Primary
Quantity 1mg
Monoclonal or Polyclonal Polyclonal