Please login to your online account to display your discounted pricing

anti-GST Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600327

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600327

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2944
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Formulation phosphate Buffered Saline (PBS)
Host Species Chicken
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Immunogen Chickens were immunized with Glutathione-S- Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Cross Reactivity Non-species specific
Quantity 0.1mg
Antigen GST Epitope Tag
Format Affinity Purified
Primary or Secondary Primary