Please login to your online account to display your discounted pricing

anti-GSK-3 beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP102156

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP102156

  • / Each
 

Description & Specifications

Specifications

Antigen GSK-3 beta
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P49841
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GSK3 beta (within residues 1-100). [Swiss-Prot No. P49841]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2932
Test Specificity Human. Predicted to react with rat, mouse, zebrafish and sheep based on 100% sequence homology.
Target Species Human