Please login to your online account to display your discounted pricing

anti-GSK-3 beta, Polyclonal, Novus Biologicals™

$128.00 - $321.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GSK3 beta (within residues 1-100). [Swiss-Prot No. P49841]
Antigen GSK-3 beta
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP102156

 
novus biologicals canada
NBP102156
Unlabeled 0.1mL Each for $321.00

NBP102156SS

 
novus biologicals canada
NBP102156SS
Unlabeled 0.025mL Each for $128.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GSK3 beta (within residues 1-100). [Swiss-Prot No. P49841]
Antigen GSK-3 beta
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Human. Predicted to react with rat, mouse, zebrafish and sheep based on 100% sequence homology.
Gene Accession No. P49841
Gene ID (Entrez) 2932
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL