missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GSAP, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5]
Antigen GSAP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178376 Novus Biologicals Canada
NBP178376
0.1mL Each for $259.02
NBP178376SS Novus Biologicals Canada
NBP178376SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GSAP
Affinity Purified
RUO
A4D1B5
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5]
Polyclonal
Primary
Simple Western,Western Blot
54103
1.11mg/mL