Please login to your online account to display your discounted pricing

anti-GRP78/HSPA5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP106274

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP106274

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation PBS, 30% glycerol
Concentration 1.26mg/mL
Regulatory Status RUO
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Sheep
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Gene Accession No. P11021
Quantity 0.1mL
Gene ID (Entrez) 3309
Test Specificity Human, mouse, rat, sheep and chicken. Predicted to react with Zebrafish, S. cerevisiae, primate, bovine, and Drosophila based on 100% sequence homology.
Format Affinity Purified
Antigen GRP78/HSPA5
Conjugate Unlabeled
Immunogen Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot No. P11021]
Host Species Rabbit