Please login to your online account to display your discounted pricing

anti-GRP78/HSPA5, Polyclonal, Novus Biologicals™

$367.12 - $367.12

Specifications

Host Species Rabbit
Immunogen This antibody was made against a protein fragment from the C Terminus Region
Antigen GRP78/HSPA5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

28-220-002

 
novus biologicals canada
28220002
Unlabeled 0.05mg Each for $367.12
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was made against a protein fragment from the C Terminus Region
Antigen GRP78/HSPA5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human HSPA5.
Gene Accession No. P11021
Gene ID (Entrez) 3309
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0