Please login to your online account to display your discounted pricing

anti-GRP78/HSPA5, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot No. P11021]
Antigen GRP78/HSPA5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP106274SS

 
novus biologicals canada
NBP106274SS
Unlabeled 0.025mL Each for $144.63

NBP106274

 
novus biologicals canada
NBP106274
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot No. P11021]
Antigen GRP78/HSPA5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Test Specificity Human, mouse, rat, sheep and chicken. Predicted to react with Zebrafish, S. cerevisiae, primate, bovine, and Drosophila based on 100% sequence homology.
Gene Accession No. P11021
Gene ID (Entrez) 3309
Formulation PBS, 30% glycerol
Concentration 1.26mg/mL