missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GRK5, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350-400 of human GRK5 was used as the immunogen.
Antigen GRK5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224580 Novus Biologicals Canada
NBP224580
0.1mg Each for $255.80
NBP224580SS Novus Biologicals Canada
NBP224580SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GRK5
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, bovine and dog, and 93% in chimp and chicken.
2869
0.5mg/mL
A portion of amino acid 350-400 of human GRK5 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P34947
100μg in 200μL PBS containing 0.2% gelatin