Please login to your online account to display your discounted pricing

anti-Gremlin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056883SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056883T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O60565
Regulatory Status RUO
Gene ID (Entrez) 26585
Concentration 0.5mg/mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse (94%) and Rat (94%).
Host Species Rabbit
Format Purified
Quantity 0.025mg
Antigen Gremlin 1
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide containing amino acids QPGSRNRGRGQGRGTA (52-67) of human CKTSF1B1 (NM_013372).