Please login to your online account to display your discounted pricing

anti-Granulysin, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 from human GNLY was used as the immunogen for this antibody.
Antigen Granulysin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227101SS

 
novus biologicals canada
NBP227101SS
Unlabeled 0.025mg Each for $109.00

NBP227101

 
novus biologicals canada
NBP227101
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 from human GNLY was used as the immunogen for this antibody.
Antigen Granulysin
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this GNLY antibody is 100% homologous in human, and chimpanzee, and 81% homologous in orangutan.
Gene Accession No. P22749
Gene ID (Entrez) 10578
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL