Please login to your online account to display your discounted pricing

anti-Granulin, Polyclonal, Novus Biologicals™

$406.00 - $406.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Granulin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

26-320-00201

 
novus biologicals canada
2632000201MG
Unlabeled 0.1mg Each for $406.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Granulin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Neutralization
Applications Western Blot
Test Specificity This product is specific for Human grn. Human. primate reactivity reported in scientific literature (PMID: 24163244).
Gene Accession No. P28799
Gene ID (Entrez) 2896
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0