Please login to your online account to display your discounted pricing

anti-Gr-1/Ly-6G, Clone: RB6-8C5, R&D Systems

$139.08 - $139.08

Specifications

Antigen Gr-1/Ly-6G
Clone RB6-8C5
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MLDP6

 
r & d systems
RDSMLDP6
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen Gr-1/Ly-6G
Clone RB6-8C5
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion