Please login to your online account to display your discounted pricing

anti-Gr-1/Ly-6G, Clone: RB6-8C5, R&D Systems


Manufacturer: r & d systems  RDSMLDP6

 View more versions of this product

Catalog No. MLDP6

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Clone RB6-8C5
Quantity 1 Vial
Antigen Gr-1/Ly-6G
Cross Reactivity Mouse
Host Species Rat
Format Affinity Purified
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Isotype IgG2b