Please login to your online account to display your discounted pricing

anti-GPR83, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224515

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224515

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9NYM4
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Gene ID (Entrez) 10888
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, cat, chimpanzee and cow and 94% homologous in chicken, horse, pig, platypus, opossum, rhesus monkey, gorilla and Xenopus.
Antigen GPR83
Immunogen A portion of amino acids 200-250 of human GPR83 was used as the immunogen for this antibody.
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Primary or Secondary Primary
Format Affinity Purified
Host Species Rabbit