Please login to your online account to display your discounted pricing

anti-GPR83, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR83 was used as the immunogen for this antibody.
Antigen GPR83
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224515

 
novus biologicals canada
NBP224515
Unlabeled 0.1mg Each for $309.05

NBP224515SS

 
novus biologicals canada
NBP224515SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR83 was used as the immunogen for this antibody.
Antigen GPR83
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, cat, chimpanzee and cow and 94% homologous in chicken, horse, pig, platypus, opossum, rhesus monkey, gorilla and Xenopus.
Gene Accession No. Q9NYM4
Gene ID (Entrez) 10888
Formulation 100μg antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL