Please login to your online account to display your discounted pricing

anti-GPR64 Polyclonal, Thermo Scientific™

GPR64 Polyclonal antibody PA5-33752 from Thermo Scientific™ specifically detects GPR64 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

$506.05 - $506.05

Specifications

Antigen GPR64
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR64.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533752

 
pierce biotechnology inc
PA533752
Unlabeled 50μg Each for $506.05

PIPA533751

 
pierce biotechnology inc
PA533751
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen GPR64
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR64.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

GPR64 Polyclonal antibody PA5-33752 from Thermo Scientific™ specifically detects GPR64 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors. TRAIL/Apo2L is a new member of the TNF family and induces apoptosis of a variety of tumor cell lines. DR4 and DR5 are the recently identified functional receptors for TRAIL, and DcR1/TRID is a decoy receptor. Another member of the TRAIL receptor family was more recently identified and designated DcR2, TRAIL-R4, or TRUNDD. DcR2 has an extracellular TRAIL-binding domain but lacks intracellular death domain and does not induce apoptosis. Like DR4 and DR5, DcR2 transcript is widely expressed in normal human tissues. Overexpression of DcR2 attenuated TRAIL-induced apoptosis.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 16 amino acid peptide from C-terminus of human GPR64.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: GPR64, Epididymal protein 6, G protein-coupled receptor 64, EDDM6, G-protein coupled receptor 64, HE6, TM7LN2
Uniprot Accession: Q8IZP9
Gene Symbol: GPR64
Entrez GeneID: 10149
PA5-33397 detects CCR6/CD196 in Human samples. PA5-33397 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-33397 The immunogen is Synthetic 18 amino acid peptide from 2nd cytoplasmic domain of human CCR6..