Please login to your online account to display your discounted pricing

anti-GPR39, Polyclonal, Novus Biologicals™

$182.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR39 protein (within residues 150-200). [Swiss-Prot O43194]
Antigen GPR39
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLS139

 
novus biologicals canada
NLS139
Unlabeled 0.05mL Each for $329.00

NLS139SS

 
novus biologicals canada
NLS139SS
Unlabeled 0.025mL Each for $182.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR39 protein (within residues 150-200). [Swiss-Prot O43194]
Antigen GPR39
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. O43194
Gene ID (Entrez) 2863
Formulation PBS, 30% glycerol
Concentration 1.1mg/mL