Please login to your online account to display your discounted pricing

anti-GPR35, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224640

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224640

  • / Each
 

Description & Specifications

Specifications

Antigen GPR35
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 275-325 of human G Protein-Coupled Receptor GPR35 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2859
Target Species Human
Target Species Murine