missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GPR18, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR18 was used as the immunogen.
Antigen GPR18
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224918 Novus Biologicals Canada
NBP224918
0.1mg Each for $307.00
NBP224918SS Novus Biologicals Canada
NBP224918SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GPR18
Affinity Purified
RUO
2841
0.5mg/mL
A portion of amino acids 1-50 of human GPR18 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
100μg antibody in 200μL PBS containing 0.05% BSA