Please login to your online account to display your discounted pricing

anti-GPR155, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP150626SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP150626SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. Q7Z3F1
Primary or Secondary Primary
Concentration 0.85mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Gene ID (Entrez) 151556
Formulation PBS, 30% glycerol
Host Species Rabbit
Cross Reactivity Human
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 800-900) [UniProt Q7Z3F1]
Antigen GPR155