Please login to your online account to display your discounted pricing

anti-GPR155, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP150625

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP150625

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Conjugate Unlabeled
Test Specificity Human. Immunogen sequence has 86% identity to mouse.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 500-600) [UniProt Q7Z3F1]
Primary or Secondary Primary
Quantity 0.1mL
Gene ID (Entrez) 151556
Gene Accession No. Q7Z3F1
Concentration 1.06mg/mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Antigen GPR155
Format Affinity Purified
Host Species Rabbit
Cross Reactivity Human