Please login to your online account to display your discounted pricing

anti-GPR155, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 500-600) [UniProt Q7Z3F1]
Antigen GPR155
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP150625

 
novus biologicals canada
NBP150625
Unlabeled 0.1mL Each for $309.05

NBP150625SS

 
novus biologicals canada
NBP150625SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 500-600) [UniProt Q7Z3F1]
Antigen GPR155
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Immunogen sequence has 86% identity to mouse.
Gene Accession No. Q7Z3F1
Gene ID (Entrez) 151556
Formulation PBS, 30% glycerol
Concentration 1.06mg/mL