Please login to your online account to display your discounted pricing

GPR149 Polyclonal Antibody, Invitrogen™


GPR149 Antibody, PA5-33639, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).

Manufacturer: pierce biotechnology inc  PA533639

Catalog No. PIPA533639

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 344758
Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Gene Accession No. Q86SP6
Species Reactivity Human
Gene Symbols GPR149
Gene Alias GPR149, PGR10, G protein-coupled receptor 149, IEDA
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Isotype IgG
Antigen GPR149
Cross Reactivity Human
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR149.
Quantity 50μg
Format Purified
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal

GPR149 Antibody, PA5-33639, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).


Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 16 amino acid peptide from C-terminus of human GPR149.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: GPR149, PGR10, G protein-coupled receptor 149, IEDA
Uniprot Accession: Q86SP6
Entrez GeneID: 344758
Gene Symbol: GPR149
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Rabbit (94%) Marmoset (81%).