Please login to your online account to display your discounted pricing

anti-GPR149 Polyclonal, Thermo Scientific™

GPR149 Polyclonal antibody PA5-33639 from Thermo Scientific™ specifically detects GPR149 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

$506.05 - $506.05

Specifications

Antigen GPR149
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR149.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533640

 
pierce biotechnology inc
PA533640
Unlabeled 50μg Each for $506.05

PIPA533639

 
pierce biotechnology inc
PA533639
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen GPR149
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR149.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

GPR149 Polyclonal antibody PA5-33639 from Thermo Scientific™ specifically detects GPR149 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

TRH-R is a 398 amino acid integral membrane protein with six transmembrane domains and belongs to G-protein coupled receptor 1 family. It acts as a receptor for the signaling hormone TRH and activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon binding with TRH. Reports suggest that TRH-R is a neurotransmitter and neuro regulator that controls dimerization of the TRH receptor and potentiates hormone-induced receptor trafficking. It is known to widely express in the thyrotroph cells of the anterior pituitary.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 16 amino acid peptide from C-terminus of human GPR149.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: GPR149, PGR10, G protein-coupled receptor 149, IEDA
Uniprot Accession: Q86SP6
Gene Symbol: GPR149
Entrez GeneID: 344758
PA5-34277 detects TRH Receptor/TRHR in Human samples. PA5-34277 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-34277 The immunogen is Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TRH Receptor..