Please login to your online account to display your discounted pricing

GPR149 Polyclonal Antibody, Invitrogen™


GPR149 Antibody, PA5-33640, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).

Manufacturer: pierce biotechnology inc  PA533640

Catalog No. PIPA533640

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 344758
Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Gene Accession No. Q86SP6
Species Reactivity Human
Gene Alias GPR149, PGR10, G protein-coupled receptor 149, IEDA
Gene Symbols GPR149
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Format Purified
Isotype IgG
Immunogen Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GPR149.
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Quantity 50μg
Antigen GPR149

GPR149 Antibody, PA5-33640, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).


Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GPR149.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: GPR149, PGR10, G protein-coupled receptor 149, IEDA
Uniprot Accession: Q86SP6
Entrez GeneID: 344758
Gene Symbol: GPR149
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bat, Pig (100%) Rabbit (93%) Elephant, Bovine (87%).