Please login to your online account to display your discounted pricing

anti-GPR119, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NLS548

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NLS548

  • / Each
 

Description & Specifications

Specifications

Antigen GPR119
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q8TDV5
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR119 protein (within residues 150-250). [Swiss-Prot Q8TDV5]
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 139760
Test Specificity Human and mouse. Immunogen sequence has 88% identity to rat.
Target Species Human
Target Species Murine